Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnpy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016251.2 | NP_001016251.1 | 4 - 5 | 472 | 81 | |
GenBank mRNA | BC166940.1 | - | 4 - 5 | 697 | 81 | |
BC170706.1 | - | 3 - 4 | 437 | 81 | ||
BC170708.1 | - | 3 - 4 | 437 | 81 | ||
CR761752.2 | - | 4 - 5 | 472 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |