Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016252.2 | NP_001016252.1 | 4 - 5 | 359 | 101 | |
GenBank mRNA | BC121540.1 | - | 4 - 5 | 373 | 101 | |
CR761750.2 | - | 4 - 5 | 359 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |