Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cby1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016253.2 | NP_001016253.1 | 4 - 5 | 367 | 77 | |
XM_012961077.2 | XP_012816531.1 | 3 - 4 | 339 | 77 | ||
XM_012961078.2 | XP_012816532.1 | 4 - 5 | 437 | 77 | ||
GenBank mRNA | BC167320.1 | - | 4 - 5 | 428 | 77 | |
BC170579.1 | - | 4 - 5 | 349 | 77 | ||
BC170581.1 | - | 4 - 5 | 349 | 77 | ||
CR761748.2 | - | 4 - 5 | 367 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |