Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdr39u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016261.2 | NP_001016261.1 | 4 - 5 | 374 | 93 | |
GenBank mRNA | BC157797.1 | - | 2 - 3 | 708 | 93 | |
BC170744.1 | - | 4 - 5 | 351 | 93 | ||
BC170746.1 | - | 4 - 5 | 351 | 93 | ||
CR761700.2 | - | 4 - 5 | 374 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |