Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016297.2 | NP_001016297.1 | 2 - 3 | 175 | 110 | |
GenBank mRNA | BC157179.1 | - | 2 - 3 | 173 | 110 | |
CR761535.2 | - | 2 - 3 | 175 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |