Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016397.2 | NP_001016397.1 | 3 - 4 | 540 | 93 | |
GenBank mRNA | BC170714.1 | - | 3 - 4 | 538 | 93 | |
BC170716.1 | - | 3 - 4 | 538 | 93 | ||
CR761158.2 | - | 3 - 4 | 540 | 93 | ||
CR761349.2 | - | 3 - 4 | 522 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |