Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016403.2 | NP_001016403.1 | 1 - 2 | 72 | 91 | |
XM_012953065.1 | XP_012808519.1 | 2 - 3 | 319 | 91 | ||
XM_012953066.2 | XP_012808520.1 | 2 - 3 | 298 | 91 | ||
XM_012953067.1 | XP_012808521.1 | 2 - 3 | 254 | 91 | ||
XM_012953069.2 | XP_012808523.1 | 2 - 3 | 233 | 91 | ||
GenBank mRNA | BC170934.1 | - | 1 - 2 | 70 | 91 | |
BC170936.1 | - | 1 - 2 | 70 | 91 | ||
CR761141.2 | - | 1 - 2 | 72 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |