Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prkaca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016403.2 | NP_001016403.1 | 4 - 5 | 361 | 65 | |
XM_012953064.1 | XP_012808518.1 | 4 - 5 | 749 | 65 | ||
XM_012953065.1 | XP_012808519.1 | 5 - 6 | 608 | 65 | ||
XM_012953066.2 | XP_012808520.1 | 5 - 6 | 587 | 65 | ||
XM_012953067.1 | XP_012808521.1 | 5 - 6 | 543 | 65 | ||
XM_012953069.2 | XP_012808523.1 | 5 - 6 | 522 | 65 | ||
XM_018092085.1 | XP_017947574.1 | 4 - 5 | 403 | 65 | ||
GenBank mRNA | BC170934.1 | - | 4 - 5 | 359 | 65 | |
BC170936.1 | - | 4 - 5 | 359 | 65 | ||
CR761141.2 | - | 4 - 5 | 361 | 65 | ||
FN562397.1 | - | 4 - 5 | 431 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |