Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mreg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016432.2 | NP_001016432.1 | 6 - 7 | 672 | 69 | |
XM_012970420.1 | XP_012825874.1 | 16 - 17 | 2057 | 69 | ||
XM_012970423.2 | XP_012825877.1 | 7 - 8 | 847 | 69 | ||
XM_018097106.1 | XP_017952595.1 | 6 - 7 | 1037 | 69 | ||
GenBank mRNA | BC122004.1 | - | 6 - 7 | 605 | 69 | |
CR761048.2 | - | 6 - 7 | 672 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |