Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016433.2 | NP_001016433.1 | 3 - 4 | 977 | 100 | |
XM_012968410.2 | XP_012823864.1 | 3 - 4 | 1143 | 100 | ||
GenBank mRNA | BC170942.1 | - | 3 - 4 | 934 | 100 | |
BC170944.1 | - | 3 - 4 | 934 | 100 | ||
CR761045.2 | - | 3 - 4 | 977 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |