Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016437.2 | NP_001016437.1 | 2 - 3 | 389 | 92 | |
GenBank mRNA | BC170899.1 | - | 2 - 3 | 320 | 92 | |
BC170903.1 | - | 2 - 3 | 320 | 92 | ||
CR761033.2 | - | 2 - 3 | 389 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |