Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bet1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016442.2 | NP_001016442.1 | 2 - 3 | 252 | 91 | |
GenBank mRNA | BC167323.1 | - | 2 - 3 | 244 | 91 | |
BC170583.1 | - | 2 - 3 | 250 | 91 | ||
BC170611.1 | - | 2 - 3 | 250 | 91 | ||
CR761023.2 | - | 2 - 3 | 252 | 91 |
Gene Symbol: LOC101730635 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004919954.2 | XP_004920011.1 | 2 - 3 | 317 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |