Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: commd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016464.2 | NP_001016464.1 | 5 - 6 | 439 | 77 | |
XM_012956969.2 | XP_012812423.1 | 6 - 7 | 567 | 77 | ||
XM_012956971.2 | XP_012812425.1 | 6 - 7 | 570 | 77 | ||
GenBank mRNA | BC171118.1 | - | 5 - 6 | 418 | 77 | |
CR760938.2 | - | 5 - 6 | 439 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |