Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016473.2 | NP_001016473.1 | 5 - 6 | 497 | 76 | |
GenBank mRNA | BC157231.1 | - | 6 - 7 | 699 | 76 | |
CR760906.2 | - | 5 - 6 | 497 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |