Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tekt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016475.2 | NP_001016475.1 | 5 - 6 | 959 | 79 | |
GenBank mRNA | BC135257.1 | - | 5 - 6 | 963 | 79 | |
CR760902.2 | - | 5 - 6 | 959 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |