Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tulp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016482.2 | NP_001016482.1 | 2 - 3 | 175 | 118 | |
GenBank mRNA | CR760884.2 | - | 2 - 3 | 175 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Xenopus tropicalis Probe spans exons |