Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmem184c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016532.2 | NP_001016532.1 | 4 - 5 | 612 | 57 | |
GenBank mRNA | BC157533.1 | - | 4 - 5 | 640 | 57 | |
CR926175.2 | - | 4 - 5 | 612 | 57 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |