Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016537.2 | NP_001016537.1 | 2 - 3 | 424 | 88 | |
XM_012956976.2 | XP_012812430.1 | 3 - 4 | 554 | 88 | ||
XM_012956977.2 | XP_012812431.1 | 3 - 4 | 522 | 88 | ||
XM_012956978.2 | XP_012812432.1 | 3 - 4 | 553 | 88 | ||
XM_012956979.2 | XP_012812433.1 | 3 - 4 | 533 | 88 | ||
XM_012956980.2 | XP_012812434.1 | 3 - 4 | 504 | 88 | ||
GenBank mRNA | BC091032.1 | - | 2 - 3 | 424 | 88 | |
CR760779.2 | - | 2 - 3 | 424 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |