Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aldh3a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016537.2 | NP_001016537.1 | 9 - 10 | 1484 | 89 | |
XM_012956978.2 | XP_012812432.1 | 10 - 11 | 1613 | 89 | ||
GenBank mRNA | BC091032.1 | - | 9 - 10 | 1484 | 89 | |
CR760779.2 | - | 9 - 10 | 1484 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |