Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cnot8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016578.2 | NP_001016578.1 | 5 - 6 | 791 | 72 | |
GenBank mRNA | BC170730.1 | - | 4 - 5 | 717 | 72 | |
BC170734.1 | - | 4 - 5 | 717 | 72 | ||
CR760639.2 | - | 5 - 6 | 791 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |