Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016580.2 | NP_001016580.1 | 1 - 2 | 269 | 59 | |
GenBank mRNA | BC170771.1 | - | 1 - 2 | 257 | 59 | |
BC170773.1 | - | 1 - 2 | 257 | 59 | ||
CR760633.2 | - | 1 - 2 | 269 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |