Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ung | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016580.2 | NP_001016580.1 | 2 - 3 | 479 | 76 | |
GenBank mRNA | BC167353.1 | - | 1 - 2 | 427 | 76 | |
BC170771.1 | - | 2 - 3 | 467 | 76 | ||
BC170773.1 | - | 2 - 3 | 467 | 76 | ||
CR760633.2 | - | 2 - 3 | 479 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |