Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nudt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016613.2 | NP_001016613.1 | 3 - 4 | 169 | 96 | |
XM_012959033.2 | XP_012814487.1 | 3 - 4 | 207 | 96 | ||
XM_018092088.1 | XP_017947577.1 | 4 - 5 | 255 | 96 | ||
GenBank mRNA | BC170594.1 | - | 2 - 3 | 159 | 96 | |
BC170598.1 | - | 2 - 3 | 159 | 96 | ||
CR760517.2 | - | 3 - 4 | 169 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |