Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sap30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016618.2 | NP_001016618.1 | 3 - 4 | 530 | 84 | |
GenBank mRNA | BC168448.1 | - | 2 - 3 | 459 | 84 | |
BC170580.1 | - | 3 - 4 | 511 | 84 | ||
BC170582.1 | - | 3 - 4 | 511 | 84 | ||
CR760505.2 | - | 3 - 4 | 530 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |