Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: agr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016627.3 | NP_001016627.1 | 3 - 4 | 239 | 73 | |
XM_012964714.2 | XP_012820168.1 | 4 - 5 | 346 | 73 | ||
GenBank mRNA | CR760462.2 | - | 3 - 4 | 241 | 73 | |
CR942718.2 | - | 1648 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Xenopus tropicalis Probe spans exons |