Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: supt4h1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016679.2 | NP_001016679.1 | 4 - 5 | 294 | 67 | |
GenBank mRNA | CR760209.2 | - | 4 - 5 | 294 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Xenopus tropicalis Probe spans exons |