Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hesx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016712.2 | NP_001016712.1 | 1 - 2 | 171 | 60 | |
GenBank mRNA | BC159020.1 | - | 1 - 2 | 171 | 60 | |
CR760108.2 | - | 1 - 2 | 171 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |