Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rps10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016718.3 | NP_001016718.1 | 2 - 3 | 323 | 103 | |
XM_012962754.2 | XP_012818208.1 | 3 - 4 | 429 | 103 | ||
XM_012962759.1 | XP_012818213.1 | 3 - 4 | 371 | 103 | ||
XM_018091267.1 | XP_017946756.1 | 3 - 4 | 475 | 103 | ||
GenBank mRNA | BC156036.1 | - | 2 - 3 | 331 | 103 | |
CR760085.2 | - | 2 - 3 | 325 | 103 | ||
CR760262.2 | - | 1797 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |