Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: derl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016723.2 | NP_001016723.1 | 6 - 7 | 566 | 73 | |
GenBank mRNA | CR760062.2 | - | 6 - 7 | 566 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |