Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016768.2 | NP_001016768.1 | 6 - 7 | 925 | 80 | |
XM_012961123.2 | XP_012816577.1 | 6 - 7 | 960 | 80 | ||
XM_012961124.1 | XP_012816578.1 | 6 - 7 | 984 | 80 | ||
XM_012961125.2 | XP_012816579.1 | 6 - 7 | 929 | 80 | ||
GenBank mRNA | BC158145.1 | - | 6 - 7 | 990 | 80 | |
CR855743.2 | - | 6 - 7 | 925 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |