Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mat2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016828.3 | NP_001016828.2 | 4 - 5 | 580 | 62 | |
GenBank mRNA | BC093462.1 | - | 4 - 5 | 580 | 62 | |
CR855516.2 | - | 4 - 5 | 599 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |