Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: washc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016854.2 | NP_001016854.1 | 3 - 4 | 460 | 60 | |
XM_012959046.2 | XP_012814500.1 | 4 - 5 | 560 | 60 | ||
XM_012959047.1 | XP_012814501.1 | 4 - 5 | 474 | 60 | ||
XM_012959048.2 | XP_012814502.1 | 4 - 5 | 560 | 60 | ||
GenBank mRNA | BC123023.1 | - | 3 - 4 | 428 | 60 | |
CR855419.2 | - | 3 - 4 | 460 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |