Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ciao1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016887.2 | NP_001016887.1 | 3 - 4 | 467 | 71 | |
XM_012959054.2 | XP_012814508.1 | 3 - 4 | 462 | 71 | ||
XM_012959056.2 | XP_012814510.1 | 3 - 4 | 394 | 71 | ||
XM_012959057.2 | XP_012814511.1 | 3 - 4 | 387 | 71 | ||
GenBank mRNA | BC127279.1 | - | 3 - 4 | 461 | 71 | |
CR848572.2 | - | 3 - 4 | 467 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |