Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001016906.2 | NP_001016906.1 | 5 - 6 | 797 | 86 | |
XM_012968448.2 | XP_012823902.1 | 5 - 6 | 884 | 86 | ||
GenBank mRNA | CR848507.2 | - | 5 - 6 | 797 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |