Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017041.2 | NP_001017041.1 | 4 - 5 | 387 | 82 | |
GenBank mRNA | BC158409.1 | - | 5 - 6 | 396 | 82 | |
CR761902.2 | - | 4 - 5 | 387 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |