Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mapk12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017080.2 | NP_001017080.1 | 4 - 5 | 795 | 91 | |
XM_018092105.1 | XP_017947594.1 | 5 - 6 | 978 | 91 | ||
GenBank mRNA | BC131830.1 | - | 5 - 6 | 821 | 91 | |
BC167997.1 | - | 4 - 5 | 739 | 91 | ||
CR761485.2 | - | 4 - 5 | 795 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |