Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017100.2 | NP_001017100.1 | 7 - 8 | 653 | 99 | |
XM_012957031.2 | XP_012812485.1 | 8 - 9 | 914 | 99 | ||
GenBank mRNA | CR761298.2 | - | 7 - 8 | 653 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |