Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrpl35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017111.2 | NP_001017111.1 | 3 - 4 | 294 | 90 | |
GenBank mRNA | BC153685.1 | - | 3 - 4 | 521 | 90 | |
CR761212.2 | - | 3 - 4 | 294 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |