Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uchl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017120.2 | NP_001017120.1 | 6 - 7 | 784 | 82 | |
GenBank mRNA | BC135703.1 | - | 6 - 7 | 530 | 82 | |
CR761162.2 | - | 6 - 7 | 784 | 82 | ||
CR761882.2 | - | 4 - 5 | 131 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |