Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: taldo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017131.2 | NP_001017131.1 | 1 - 2 | 272 | 75 | |
GenBank mRNA | CR761089.2 | - | 1 - 2 | 272 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Xenopus tropicalis Probe spans exons |