Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: yju2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017176.2 | NP_001017176.1 | 6 - 7 | 769 | 78 | |
XM_012963057.2 | XP_012818511.1 | 6 - 7 | 870 | 78 | ||
GenBank mRNA | BC157156.1 | - | 6 - 7 | 761 | 78 | |
CR760706.2 | - | 6 - 7 | 769 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Xenopus tropicalis Probe spans exons |