Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctdnep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017177.2 | NP_001017177.1 | 454 | 59 | ||
GenBank mRNA | CR760700.2 | - | 454 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Xenopus tropicalis |