Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scamp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017190.2 | NP_001017190.1 | 1 - 2 | 74 | 86 | |
GenBank mRNA | BC167666.1 | - | 1 - 2 | 95 | 86 | |
CR760611.2 | - | 1 - 2 | 74 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |