Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017191.2 | NP_001017191.1 | 1 - 2 | 147 | 80 | |
GenBank mRNA | BC121552.1 | - | 1 - 2 | 66 | 80 | |
CR760599.2 | - | 1 - 2 | 147 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |