Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: klhl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017289.2 | NP_001017289.1 | 3 - 4 | 1031 | 60 | |
XM_018093771.1 | XP_017949260.1 | 4 - 5 | 2507 | 60 | ||
XM_018093773.1 | XP_017949262.1 | 3 - 4 | 820 | 60 | ||
GenBank mRNA | BC167576.1 | - | 4 - 5 | 1074 | 60 | |
CR855674.2 | - | 3 - 4 | 1031 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |