Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017319.2 | NP_001017319.1 | 4 - 5 | 893 | 113 | |
XM_012961199.2 | XP_012816653.1 | 4 - 5 | 910 | 113 | ||
GenBank mRNA | BC167905.1 | - | 4 - 5 | 932 | 113 | |
CR855411.2 | - | 4 - 5 | 893 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |