Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mhc1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037273.1 | NP_001032350.2 | 4 - 5 | 884 | 62 | |
GenBank mRNA | BC089748.1 | - | 4 - 5 | 884 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Xenopus tropicalis Probe spans exons |