Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039740.1 | NP_001034829.1 | 10 - 11 | 1159 | 81 | |
XM_012961021.1 | XP_012816475.1 | 11 - 12 | 1285 | 81 | ||
XM_012961023.1 | XP_012816477.1 | 11 - 12 | 1287 | 81 | ||
XM_018093032.1 | XP_017948521.1 | 11 - 12 | 1285 | 81 | ||
GenBank mRNA | BC089700.1 | - | 10 - 11 | 1136 | 81 | |
BC167302.1 | - | 10 - 11 | 1141 | 81 | ||
BC170700.1 | - | 10 - 11 | 1136 | 81 | ||
BC170702.1 | - | 10 - 11 | 1136 | 81 | ||
CR762362.2 | - | 10 - 11 | 1159 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Xenopus tropicalis Probe spans exons |