Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctu2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039740.1 | NP_001034829.1 | 6 - 7 | 499 | 86 | |
XM_012961021.1 | XP_012816475.1 | 7 - 8 | 86 | |||
XM_012961023.1 | XP_012816477.1 | 7 - 8 | 86 | |||
XM_018093032.1 | XP_017948521.1 | 7 - 8 | 86 | |||
GenBank mRNA | BC089700.1 | - | 6 - 7 | 86 | ||
BC167302.1 | - | 6 - 7 | 481 | 86 | ||
BC170700.1 | - | 6 - 7 | 476 | 86 | ||
BC170702.1 | - | 6 - 7 | 476 | 86 | ||
CR762362.2 | - | 6 - 7 | 499 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Xenopus tropicalis Probe spans exons |