Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc5a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | CR761796.2 | - | 6 - 7 | 1023 | 98 | |
CU075779.1 | - | 6 - 7 | 1025 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Xenopus tropicalis Probe spans exons |